(R)-3-Amino-3-(3-fluorophenyl)propionic acid 97%
Leverantör: Apollo Scientific
Synonyms: (R)-3-Amino-3-(3-fluorophenyl)propanoic acid
PC530014-1G PC530014-5G
APOSPC530014-1GEA 1650 SEK
APOSPC530014-1G APOSPC530014-5G
(R)-3-Amino-3-(3-fluorophenyl)propionic acid 97%
(R)-3-Amino-3-(3-fluorophenyl)propionic acid
Formel: C₉H₁₀FNO₂
MW: 183.18 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD04113656
CAS nummer: 723284-81-9