4-Bromo-2,6-dichlorobenzotrifluoride 95%
Leverantör: Apollo Scientific
PC52781-1G PC52781-250MG PC52781-5G
APOSPC52781-1GEA 8225 SEK
APOSPC52781-1G APOSPC52781-250MG APOSPC52781-5G
4-Bromo-2,6-dichlorobenzotrifluoride 95%
4-Bromo-2,6-dichlorobenzotrifluoride
Formel: C₇H₂BrCl₂F₃
MW: 293,896 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD32659906
CAS nummer: 2169186-20-1