3-Chloro-4,5-difluorophenol 95%
Leverantör: Apollo Scientific
PC520897-1G PC520897-250MG PC520897-5G
APOSPC520897-1GEA 3325 SEK
APOSPC520897-1G APOSPC520897-250MG APOSPC520897-5G
3-Chloro-4,5-difluorophenol 95%
3-Chloro-4,5-difluorophenol
Formel: C₆H₃ClF₂O
MW: 164,537 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18415631
CAS nummer: 1261472-63-2