4-Fluoro-5-iodo-2-methylphenyl trifluoromethanesulphonate
Leverantör: Apollo Scientific
Fara
PC52088-1G PC52088-250MG PC52088-5G
APOSPC52088-1GEA 4800 SEK
APOSPC52088-1G APOSPC52088-250MG APOSPC52088-5G
4-Fluoro-5-iodo-2-methylphenyl trifluoromethanesulphonate
4-Fluoro-5-iodo-2-methylphenyl trifluoromethanesulphonate
Formel: C₈H₅F₄IO₃S
MW: 384.087 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28897193
CAS nummer: 1935368-66-3
UN: 3265
ADR: 8 III