[2-(Trifluoromethyl)pyrimidin-5-yl]methanol 95%
Leverantör: Apollo Scientific
Synonyms: (2-(Trifluoromethyl)pyrimidin-5-yl)methanol
PC520809-250MG PC520809-500MG
APOSPC520809-250MGEA 10550 SEK
APOSPC520809-250MG APOSPC520809-500MG
[2-(Trifluoromethyl)pyrimidin-5-yl]methanol 95%
[2-(Trifluoromethyl)pyrimidin-5-yl]methanol
Formel: C₆H₅F₃N₂O
MW: 178,112 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09863203
CAS nummer: 608515-90-8