1-Bromo-4,5-difluoro-2-(trifluoromethoxy)benzene 95%
Leverantör: Apollo Scientific
PC520767-1G PC520767-500MG
APOSPC520767-1GEA 4800 SEK
APOSPC520767-1G APOSPC520767-500MG
1-Bromo-4,5-difluoro-2-(trifluoromethoxy)benzene 95%
1-Bromo-4,5-difluoro-2-(trifluoromethoxy)benzene
Formel: C₇H₂BrF₅O
MW: 276.986 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD22580797
CAS nummer: 866632-64-6