N-[1-(2-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride 92%
Leverantör: Apollo Scientific
Fara
PC520402-100MG PC520402-1G PC520402-250MG
APOSPC520402-100MGEA 175 SEK
APOSPC520402-100MG APOSPC520402-1G APOSPC520402-250MG
N-[1-(2-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride 92%
N-[1-(2-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Formel: C₁₁H₁₂FN·ClH
MW: 213,679 g/mol
Storage Temperature: Frys
MDL Number: MFCD31745724
CAS nummer: 2379918-37-1
UN: 2811
ADR: 6.1,III