N-[1-(3-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride, Technical Grade
Leverantör: Apollo Scientific
Fara
PC520401-100MG PC520401-1G PC520401-250MG
APOSPC520401-100MGEA 175 SEK
APOSPC520401-100MG APOSPC520401-1G APOSPC520401-250MG
N-[1-(3-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride, Technical Grade
N-[1-(3-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Formel: C₁₁H₁₂FN·ClH
MW: 213,679 g/mol
Storage Temperature: Frys
MDL Number: MFCD31745723
CAS nummer: 2379918-46-2
UN: 2811
ADR: 6.1,III