5-Bromo-3-fluorobenzene-1,2-diamine 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Bromo-1,2-diamino-3-fluorobenzene
PC5177-1G PC5177-250MG PC5177-5G
APOSPC5177-1GEA 740 SEK
APOSPC5177-1G APOSPC5177-250MG APOSPC5177-5G
5-Bromo-3-fluorobenzene-1,2-diamine 97%
5-Bromo-3-fluorobenzene-1,2-diamine
Formel: C₆H₆BrFN₂
MW: 205,028 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03094270
CAS nummer: 517920-69-3