4-(Aminomethyl)-2-(difluoromethyl)pyridine hydrochloride
Leverantör: Apollo Scientific
Varning
Synonyms: [2-(Difluoromethyl)pyridin-4-yl]methylamine hydrochloride, 2-(Difluoromethyl)isonicotinylamine hydrochloride
PC51208-1G PC51208-250MG
APOSPC51208-1GEA 6950 SEK
APOSPC51208-1G APOSPC51208-250MG
4-(Aminomethyl)-2-(difluoromethyl)pyridine hydrochloride
4-(Aminomethyl)-2-(difluoromethyl)pyridine hydrochloride
Formel: C₇H₈F₂N₂·ClH
MW: 194,61 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD23701110
CAS nummer: 1428532-89-1