2-(Difluoromethyl)isonicotinic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 2-(Difluoromethyl)pyridine-4-carboxylic acid
PC51206-250MG PC51206-50MG
APOSPC51206-250MGEA 2750 SEK
APOSPC51206-250MG APOSPC51206-50MG
2-(Difluoromethyl)isonicotinic acid
2-(Difluoromethyl)isonicotinic acid
Formel: C₇H₅F₂NO₂
MW: 173.117 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18256212
CAS nummer: 1256818-14-0