2,4-Difluoro-3-(trifluoromethyl)benzoic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 3-Carboxy-2,6-difluorobenzotrifluoride, α,α,α,2,4-Pentafluoro-m-toluic acid
PC51183-1G PC51183-250MG PC51183-5G
APOSPC51183-1GEA 4275 SEK
APOSPC51183-1G APOSPC51183-250MG APOSPC51183-5G
2,4-Difluoro-3-(trifluoromethyl)benzoic acid 98%
2,4-Difluoro-3-(trifluoromethyl)benzoic acid
Formel: C₈H₃F₅O₂
MW: 226,1 g/mol
Smältpunkt: 110 °C
Storage Temperature: Omgivande
MDL Number: MFCD12827981
CAS nummer: 157337-81-0