4-Fluoro-3-iodonitrobenzene
Leverantör: Apollo Scientific
Varning
Synonyms: 1-Fluoro-2-iodo-4-nitrobenzene
PC51098-1G PC51098-5G PC51098-25G
APOSPC51098-1GEA 515 SEK
APOSPC51098-1G APOSPC51098-5G APOSPC51098-25G
4-Fluoro-3-iodonitrobenzene
4-Fluoro-3-iodonitrobenzene
Formel: C₆H₃FINO₂
MW: 266,996 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08461002
CAS nummer: 177363-10-9