1,1,1,3,3,3-Hexafluoro-2-phenylprop-2-yl methacrylate
Leverantör: Apollo Scientific
Synonyms: 1,1,1,3,3,3-Hexafluoro-2-phenylprop-2-yl 2-methylprop-2-enoate
PC51056-1G PC51056-5G
APOSPC51056-1GEA 1350 SEK
APOSPC51056-1G APOSPC51056-5G
1,1,1,3,3,3-Hexafluoro-2-phenylprop-2-yl methacrylate
1,1,1,3,3,3-Hexafluoro-2-phenylprop-2-yl methacrylate
Formel: C₁₃H₁₀F₆O₂
MW: 312.208 g/mol
Kokpunkt: 128 °C (5 mm Hg)
Storage Temperature: Omgivande
MDL Number: MFCD19442078
CAS nummer: 130540-87-3