4-Chloro-2,6-difluoroanisole
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Chloro-1,3-difluoro-2-methoxybenzene, 4-Chloro-2,6-difluorophenyl methyl ether
PC51046-1G PC51046-5G PC51046-25G
APOSPC51046-1GEA 755 SEK
APOSPC51046-1G APOSPC51046-5G APOSPC51046-25G
4-Chloro-2,6-difluoroanisole
4-Chloro-2,6-difluoroanisole
Formel: C₇H₅ClF₂O
MW: 178,564 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD13193248
CAS nummer: 170572-51-7