Methyl 4-morpholino-2-(trifluoroacetamido)benzoate
Leverantör: Apollo Scientific
PC510245-1G PC510245-5G
APOSPC510245-1GEA 5525 SEK
APOSPC510245-1G APOSPC510245-5G
Methyl 4-morpholino-2-(trifluoroacetamido)benzoate
Methyl 4-morpholino-2-(trifluoroacetamido)benzoate
Formel: C₁₄H₁₅F₃N₂O₄
MW: 332,275 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD14585011
CAS nummer: 404010-73-7