6-Bromo-5-fluoro-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole
Leverantör: Apollo Scientific
Varning
Synonyms: 2-(6-Bromo-5-fluoro-1H-indazol-1-yl)tetrahydro-2H-pyran, 2-(6-Bromo-5-fluoro-1H-indazol-1-yl)oxane
PC51017-100MG PC51017-250MG
APOSPC51017-100MGEA 2100 SEK
APOSPC51017-100MG APOSPC51017-250MG
6-Bromo-5-fluoro-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole
6-Bromo-5-fluoro-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole
Formel: C₁₂H₁₂BrFN₂O
MW: 299.139 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18384869
CAS nummer: 1286734-76-6