5-Fluoro-2-(methylsulphonamido)benzoic acid
Leverantör: Apollo Scientific
PC510054-1G PC510054-5G
APOSPC510054-1GEA 3250 SEK
APOSPC510054-1G APOSPC510054-5G
5-Fluoro-2-(methylsulphonamido)benzoic acid
5-Fluoro-2-(methylsulphonamido)benzoic acid
Formel: C₈H₈FNO₄S
MW: 233,217 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09949279
CAS nummer: 1016788-23-0