2-[4-Hydroxy-6-(trifluoromethyl)-2-pyrimidinylthio]acetic acid
Leverantör: Apollo Scientific
PC510037-25G PC510037-5G
APOSPC510037-25GEA 14350 SEK
APOSPC510037-25G APOSPC510037-5G
2-[4-Hydroxy-6-(trifluoromethyl)-2-pyrimidinylthio]acetic acid
2-[4-Hydroxy-6-(trifluoromethyl)-2-pyrimidinylthio]acetic acid
Formel: C₇H₅F₃N₂O₃S
MW: 254.186 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00665841
CAS nummer: 836-12-4