Ethyl cis-2-(4-Fluorobenzylamino)cyclopentanecarboxylate
Leverantör: Apollo Scientific
PC510024-1G PC510024-5G
APOSPC510024-1GEA 3025 SEK
APOSPC510024-1G APOSPC510024-5G
Ethyl cis-2-(4-Fluorobenzylamino)cyclopentanecarboxylate
Ethyl cis-2-(4-Fluorobenzylamino)cyclopentanecarboxylate
Formel: C₁₅H₂₀FNO₂
MW: 265,323 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18651690
CAS nummer: 1033755-81-5