Methyl 2-Amino-5-fluoro-4-morpholinobenzoate
Leverantör: Apollo Scientific
PC510002-1G PC510002-5G
APOSPC510002-1GEA 2300 SEK
APOSPC510002-1G APOSPC510002-5G
Methyl 2-Amino-5-fluoro-4-morpholinobenzoate
Methyl 2-Amino-5-fluoro-4-morpholinobenzoate
Formel: C₁₂H₁₅FN₂O₃
MW: 254,258 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11656806
CAS nummer: 864292-15-9