1-Bromo-2,3,5-trifluoro-4-iodobenzene 95%
Leverantör: Apollo Scientific
PC50549-1G PC50549-250MG
APOSPC50549-1GEA 7450 SEK
APOSPC50549-1G APOSPC50549-250MG
1-Bromo-2,3,5-trifluoro-4-iodobenzene 95%
1-Bromo-2,3,5-trifluoro-4-iodobenzene
Formel: C₆HBrF₃I
MW: 336,876 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD32669845
CAS nummer: 2514941-97-8