2-Chloro-3,6-difluorophenetole 98%
Leverantör: Apollo Scientific
Synonyms: 2-Chloro-3-ethoxy-1,4-difluorobenzene
PC50427-10G PC50427-1G PC50427-5G
APOSPC50427-10GEA 6675 SEK
APOSPC50427-10G APOSPC50427-1G APOSPC50427-5G
2-Chloro-3,6-difluorophenetole 98%
2-Chloro-3,6-difluorophenetole
Formel: C₈H₇ClF₂O
Storage Temperature: Omgivande
MDL Number: MFCD28750542
CAS nummer: 1807166-60-4