Ethyl 4-bromo-2,5-difluorobenzoate
Leverantör: Apollo Scientific
Varning
PC50404-1G PC50404-250MG PC50404-5G
APOSPC50404-1GEA 3025 SEK
APOSPC50404-1G APOSPC50404-250MG APOSPC50404-5G
Ethyl 4-bromo-2,5-difluorobenzoate
Ethyl 4-bromo-2,5-difluorobenzoate
Formel: C₉H₇BrF₂O₂
MW: 265,051 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD25459665
CAS nummer: 1383685-39-9