2-Bromo-5-chloro-4-fluorobenzoic acid 95%
Leverantör: Apollo Scientific
Varning
PC50346-1G PC50346-250MG PC50346-5G
APOSPC50346-1GEA 4800 SEK
APOSPC50346-1G APOSPC50346-250MG APOSPC50346-5G
2-Bromo-5-chloro-4-fluorobenzoic acid 95%
2-Bromo-5-chloro-4-fluorobenzoic acid
Formel: C₇H₃BrClFO₂
MW: 253,453 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD24108577
CAS nummer: 1519109-81-9