4-Ethynyl-2,6-difluoroaniline
Leverantör: Apollo Scientific
Varning
PC50309-1G PC50309-250MG
APOSPC50309-1GEA 9000 SEK
APOSPC50309-1G APOSPC50309-250MG
4-Ethynyl-2,6-difluoroaniline
4-Ethynyl-2,6-difluoroaniline
Formel: C₈H₅F₂N
MW: 153,129 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08703479
CAS nummer: 753501-37-0