2-Bromo-3-fluorophenylisothiocyanate
Leverantör: Apollo Scientific
Fara
Synonyms: 2-Bromo-1-fluoro-3-isothiocyanatobenzene
PC502994-1G PC502994-25G PC502994-5G
APOSPC502994-1GEA 940 SEK
APOSPC502994-1G APOSPC502994-25G APOSPC502994-5G
2-Bromo-3-fluorophenylisothiocyanate
2-Bromo-3-fluorophenylisothiocyanate
Formel: C₇H₃BrFNS
MW: 232.073 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD22557657
CAS nummer: 364364-00-1
UN: 2922
ADR: 8+6.1 III