Methyl 2-chloro-6-fluorobenzoate
Leverantör: Apollo Scientific
PC502674-1G PC502674-5G
APOSPC502674-1GEA 840 SEK
APOSPC502674-1G APOSPC502674-5G
Methyl 2-chloro-6-fluorobenzoate
Methyl 2-chloro-6-fluorobenzoate
Formel: C₈H₆ClFO₂
MW: 188,583 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02245077
CAS nummer: 151360-57-5