3-Bromo-2,4,5-trifluorobenzyl bromide 95+%
Leverantör: Apollo Scientific
Fara
PC502351-1G PC502351-250MG PC502351-5G
APOSPC502351-1GEA 14350 SEK
APOSPC502351-1G APOSPC502351-250MG APOSPC502351-5G
3-Bromo-2,4,5-trifluorobenzyl bromide 95+%
3-Bromo-2,4,5-trifluorobenzyl bromide
Formel: C₇H₃Br₂F₃
MW: 303.902 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12756732
CAS nummer: 1260903-09-0