4-Bromo-2,6-difluorostyrene 95%
Leverantör: Apollo Scientific
Varning
PC501812-1G PC501812-250MG PC501812-5G
APOSPC501812-1GEA 8225 SEK
APOSPC501812-1G APOSPC501812-250MG APOSPC501812-5G
4-Bromo-2,6-difluorostyrene 95%
4-Bromo-2,6-difluorostyrene
Formel: C₈H₅BrF₂
MW: 219,026 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD28360633
CAS nummer: 160975-98-4