2-Amino-5-(pentafluorothio)benzonitrile 95%
Leverantör: Apollo Scientific
Varning
PC501347-250MG PC501347-100MG
APOSPC501347-250MGEA 8675 SEK
APOSPC501347-250MG APOSPC501347-100MG
2-Amino-5-(pentafluorothio)benzonitrile 95%
2-Amino-5-(pentafluorothio)benzonitrile
Formel: C₇H₅F₅N₂S
MW: 244.185 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD28348604
CAS nummer: 159727-27-2