5-Bromo-4-fluoro-3-(trifluoromethyl)benzene-1,2-diamine
Leverantör: Apollo Scientific
Varning
Synonyms: 2,3-Diamino-5-bromo-6-fluorobenzotrifluoride
PC50108-1G PC50108-250MG
APOSPC50108-1GEA 5300 SEK
APOSPC50108-1G APOSPC50108-250MG
5-Bromo-4-fluoro-3-(trifluoromethyl)benzene-1,2-diamine
5-Bromo-4-fluoro-3-(trifluoromethyl)benzene-1,2-diamine
Formel: C₇H₅BrF₄N₂
MW: 273.026 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28515424
CAS nummer: 1820706-93-1