1,2-Bis(trifluoromethyl)-4-methylbenzene 95%
Leverantör: Apollo Scientific
Varning
PC50098-10G PC50098-1G PC50098-5G
APOSPC50098-10GEA 13600 SEK
APOSPC50098-10G APOSPC50098-1G APOSPC50098-5G
1,2-Bis(trifluoromethyl)-4-methylbenzene 95%
1,2-Bis(trifluoromethyl)-4-methylbenzene
Formel: C₉H₆F₆
MW: 228,134 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD14525508
CAS nummer: 716-25-6