6-(Hydroxymethyl)-2,2,3,3-tetrafluoro-1,4-benzodioxane
Leverantör: Apollo Scientific
Varning
PC500584-1G PC500584-250MG
APOSPC500584-1GEA 8475 SEK
APOSPC500584-1G APOSPC500584-250MG
6-(Hydroxymethyl)-2,2,3,3-tetrafluoro-1,4-benzodioxane
6-(Hydroxymethyl)-2,2,3,3-tetrafluoro-1,4-benzodioxane
Formel: C₉H₆F₄O₃
MW: 238,136 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28096595
CAS nummer: 444919-46-4