4'-Bromo-2'-(trifluoromethyl)acetophenone 95%

Leverantör: Apollo Scientific
Varning

Synonyms: 1-(4-Bromo-2-(trifluoromethyl)phenyl)ethanone

PC500539-1G PC500539-250MG PC500539-5G
APOSPC500539-1GEA 2450 SEK
APOSPC500539-1G APOSPC500539-250MG APOSPC500539-5G
4'-Bromo-2'-(trifluoromethyl)acetophenone 95%
4'-Bromo-2'-(trifluoromethyl)acetophenone
Formel: C₉H₆BrF₃O
MW: 267,043 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16839140
CAS nummer: 1197231-94-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR