Methyl 2-(bromomethyl)-3-fluorobenzoate
Leverantör: Apollo Scientific
Fara
Synonyms: α-Bromo-3-fluoro-o-toluate, 2-Fluoro-6-(methoxycarbonyl)benzyl bromide
PC500509-1G PC500509-250MG PC500509-5G
APOSPC500509-1GEA 3025 SEK
APOSPC500509-1G APOSPC500509-250MG APOSPC500509-5G
Methyl 2-(bromomethyl)-3-fluorobenzoate
Methyl 2-(bromomethyl)-3-fluorobenzoate
Formel: C₉H₈BrFO₂
MW: 247.061 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD11505952
CAS nummer: 142314-72-5
UN: 3265
ADR: 8 III