3-Bromo-6-chloro-2-fluoronitrobenzene
Leverantör: Apollo Scientific
PC500503-1G
PC500503-250MG
APOSPC500503-1GEA
9850
SEK
APOSPC500503-1G
APOSPC500503-250MG
3-Bromo-6-chloro-2-fluoronitrobenzene
3-Bromo-6-chloro-2-fluoronitrobenzene
Formel:
C₆H₂BrClFNO₂ MW: 254,441 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD28040517 CAS nummer: 1806970-78-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...