5-Bromo-2-fluorophenacyl bromide
Leverantör: Apollo Scientific
Fara
Synonyms: 2-Bromo-1-(5-bromo-2-fluorophenyl)ethan-1-one
PC500502-1G PC500502-250MG PC500502-5G
APOSPC500502-1GEA 4800 SEK
APOSPC500502-1G APOSPC500502-250MG APOSPC500502-5G
5-Bromo-2-fluorophenacyl bromide
5-Bromo-2-fluorophenacyl bromide
Formel: C₈H₅Br₂FO
MW: 295.931 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD23711739
CAS nummer: 1427413-65-7
UN: 3261
ADR: 8 III