3-(Bromomethyl)-2-fluorobenzoic acid
Leverantör: Apollo Scientific
Fara
Synonyms: 3-Carboxy-2-fluorobenzyl bromide, α-Bromo-2-fluoro-m-toluic acid
PC500470-1G PC500470-250MG PC500470-5G
APOSPC500470-1GEA 4500 SEK
APOSPC500470-1G APOSPC500470-250MG APOSPC500470-5G
3-(Bromomethyl)-2-fluorobenzoic acid
3-(Bromomethyl)-2-fluorobenzoic acid
Formel: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD18378583
CAS nummer: 680610-75-7
UN: 3261
ADR: 8 III