2-Chloro-4-fluoro-3-methylaniline
Leverantör: Apollo Scientific
Varning
PC500362-1G PC500362-250MG
APOSPC500362-1GEA 1725 SEK
APOSPC500362-1G APOSPC500362-250MG
2-Chloro-4-fluoro-3-methylaniline
2-Chloro-4-fluoro-3-methylaniline
Formel: C₇H₇ClFN
MW: 159,589 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD13193244
CAS nummer: 90292-63-0