2-Bromo-5-fluoro-4-nitroaniline
Leverantör: Apollo Scientific
PC500296-1G
PC500296-5G
APOSPC500296-1GEA
635
SEK
APOSPC500296-1G
APOSPC500296-5G
2-Bromo-5-fluoro-4-nitroaniline
2-Bromo-5-fluoro-4-nitroaniline
Formel:
C₆H₄BrFN₂O₂ MW: 235,011 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD22381005 CAS nummer: 952664-69-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...