2,4-Difluoro-5-nitrobenzotrifluoride
Leverantör: Apollo Scientific
Varning
Synonyms: 1,5-Difluoro-2-nitro-4-(trifluoromethyl)benzene
PC500228-1G PC500228-250MG
APOSPC500228-1GEA 1725 SEK
APOSPC500228-1G APOSPC500228-250MG
2,4-Difluoro-5-nitrobenzotrifluoride
2,4-Difluoro-5-nitrobenzotrifluoride
Formel: C₇H₂F₅NO₂
MW: 227.088 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16659985
CAS nummer: 35010-29-8