2-(2,2,2-Trifluoroethylamino)benzoic acid 95%

Leverantör: Apollo Scientific

PC499028-1G PC499028-250MG
APOSPC499028-1GEA 6750 SEK
APOSPC499028-1G APOSPC499028-250MG
2-(2,2,2-Trifluoroethylamino)benzoic acid 95%
2-(2,2,2-Trifluoroethylamino)benzoic acid
Formel: C₉H₈F₃NO₂
Storage Temperature: Omgivande
MDL Number: MFCD16692384
CAS nummer: 20939-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR