3,6-Bis(1,1-dimethylethyl)-10-phenyl-9-(2,4,6-trimethylphenyl)-acridinium tetrafluoroborate 95%
Leverantör: Apollo Scientific
Synonyms: di-tBu-Mes-Acr+BF4-, 3,6-Di-tert-butyl-9-mesityl-10-phenylacridin-10-ium tetrafluoroborate
PC49681-100MG PC49681-250MG PC49681-500MG
APOSPC49681-100MGEA 3500 SEK
APOSPC49681-100MG APOSPC49681-250MG APOSPC49681-500MG
3,6-Bis(1,1-dimethylethyl)-10-phenyl-9-(2,4,6-trimethylphenyl)-acridinium tetrafluoroborate 95%
3,6-Bis(1,1-dimethylethyl)-10-phenyl-9-(2,4,6-trimethylphenyl)-acridinium tetrafluoroborate
Formel: C₃₆H₄₀N·BF₄
MW: 573,514 g/mol
Smältpunkt: 277,2 °C
Storage Temperature: Kylskåp
MDL Number: MFCD31381908
CAS nummer: 1810004-87-5