2,2-Difluoroethyl methanesulphonate
Leverantör: Apollo Scientific
Synonyms: 2,2-Difluoroethylmethane sulphonate, 2,2-Difluoroethyl mesylate
PC49275-1G PC49275-250MG
APOSPC49275-1GEA 7725 SEK
APOSPC49275-1G APOSPC49275-250MG
2,2-Difluoroethyl methanesulphonate
2,2-Difluoroethyl methanesulphonate
Formel: C₃H₆F₂O₃S
MW: 160,14 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD19442174
CAS nummer: 163035-65-2