Methyl 3-bromo-4-chloro-2-fluorobenzoate 95%
Leverantör: Apollo Scientific
PC48936-1G PC48936-250MG
APOSPC48936-1GEA 7875 SEK
APOSPC48936-1G APOSPC48936-250MG
Methyl 3-bromo-4-chloro-2-fluorobenzoate 95%
Methyl 3-bromo-4-chloro-2-fluorobenzoate
Formel: C₈H₅BrClFO₂
MW: 267,48 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28733929
CAS nummer: 1804897-28-6