1-(8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3-yl)ethanol
Leverantör: Apollo Scientific
Varning
PC48784-1G PC48784-250MG
APOSPC48784-1GEA 1725 SEK
APOSPC48784-1G APOSPC48784-250MG
1-(8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3-yl)ethanol
1-(8-Chloro-6-(trifluoromethyl)-[1,2,4]triazolo[4,3-a]pyridin-3-yl)ethanol
Formel: C₉H₇ClF₃N₃O
MW: 265,62 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD30726053
CAS nummer: 1823182-85-9