2-Bromo-6-fluoro-3-methoxybenzaldehyde 97%
Leverantör: Apollo Scientific
PC48652-1G PC48652-5G
APOSPC48652-1GEA 2575 SEK
APOSPC48652-1G APOSPC48652-5G
2-Bromo-6-fluoro-3-methoxybenzaldehyde 97%
2-Bromo-6-fluoro-3-methoxybenzaldehyde
Formel: C₈H₆BrFO₂
MW: 233,034 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD11856027
CAS nummer: 154650-22-3