1,3'-Biazetidine bis(trifluoroacetate)
Leverantör: Apollo Scientific
Varning
PC48464-100MG PC48464-250MG
APOSPC48464-100MGEA 3450 SEK
APOSPC48464-100MG APOSPC48464-250MG
1,3'-Biazetidine bis(trifluoroacetate)
1,3'-Biazetidine bis(trifluoroacetate)
Formel: C₆H₁₂N₂·2CF₃CO₂H
MW: 340.22 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD18839202
CAS nummer: 1909335-95-0