5-Bromo-4-fluoro-2-methylanisole
Leverantör: Apollo Scientific
Varning
Synonyms: 4-Bromo-5-fluoro-2-methoxytoluene, 5-Bromo-4-fluoro-2-methylphenyl methyl ether, 1-Bromo-2-fluoro-5-methoxy-4-methylbenzene
PC48456-1G PC48456-250MG PC48456-5G PC48456-25G
APOSPC48456-1GEA 4100 SEK
APOSPC48456-1G APOSPC48456-250MG APOSPC48456-5G APOSPC48456-25G
5-Bromo-4-fluoro-2-methylanisole
5-Bromo-4-fluoro-2-methylanisole
Formel: C₈H₈BrFO
MW: 219.051 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD28096617
CAS nummer: 1785363-64-5